Moronic acid ≥96.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2610-5MG M2610-25MG
TCIAM2610-5MGEA 200 EUR
TCIAM2610-5MG TCIAM2610-25MG
Moronic acid ≥96.0% (by HPLC)
Moronic acid
Formule: C₃₀H₄₆O₃
MDL Nummer: MFCD11113464
CAS nummer: 6713-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aad77c0dcbb76075e457e757d41a767342bece4&vid=afa8bbdd83dfc2e82065618980ceac5524b5402b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) (CAD) min. 96.0 area%
Specific rotation [a]20/D +64.0 to +70.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR