3-Methylcyclopentadecanone ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Methylcyclopentadecanone, Muscone
M2611-200MG
M2611-1G
TCIAM2611-200MGEA
72.6
EUR
TCIAM2611-200MG
TCIAM2611-1G
3-Methylcyclopentadecanone ≥97.0% (by GC)
3-Methylcyclopentadecanone
Formule:
C₁₆H₃₀O Kookpunt: 95°C Smeltpunt: –15°C |
MDL Nummer:
MFCD00211114 CAS nummer: 541-91-3 EINECS: 208-795-8 Merck Index: 14,06313 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...