Mianserin hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2,3,4,10,14b-Hexahydro-2-methyldibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride

M2623-200MG M2623-1G
TCIAM2623-200MGEA 37.4 EUR
TCIAM2623-200MG TCIAM2623-1G
Mianserin hydrochloride ≥98.0% (by HPLC, titration analysis)
Mianserin hydrochloride
Formule: C₁₈H₂₀N₂
Smeltpunt: 284°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00055072
CAS nummer: 21535-47-7
EINECS: 244-426-7
Merck Index: 14,06172
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2150ba11abcac5a89e33567018545ff334cc00ec&vid=2150ba11abcac5a89e33567018545ff334cc00ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR