3-Methyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2643-200MG
M2643-1G
TCIAM2643-200MGEA
34.1
EUR
TCIAM2643-200MG
TCIAM2643-1G
3-Methyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
3-Methyl-1H-pyrazole-4-carbaldehyde
Formule:
C5H6N2O Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01313822 CAS nummer: 112758-40-4 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 112.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...