4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2647-1G
M2647-5G
TCIAM2647-1GEA
35.2
EUR
TCIAM2647-1G
TCIAM2647-5G
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formule:
C₁₁H₁₇BO₂S Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD11045430 CAS nummer: 635305-48-5 |
|
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...