2-(3-Methoxyphenyl)ethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-Methoxyphenethyl alcohol

M2658-1G M2658-5G
TCIAM2658-1GEA 38.5 EUR
TCIAM2658-1G TCIAM2658-5G
2-(3-Methoxyphenyl)ethanol ≥98.0% (by GC)
2-(3-Methoxyphenyl)ethanol
Formule: C₉H₁₂O₂
Kookpunt: 140°C
MDL Nummer: MFCD00002893
CAS nummer: 5020-41-7
EINECS: 225-705-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bec231d7c4245e3be2c02d12b660104b8e04843&vid=5bec231d7c4245e3be2c02d12b660104b8e04843&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR