N-Methoxy-N-methylbenzamide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2672-5G M2672-25G
TCIAM2672-5GEA 59.4 EUR
TCIAM2672-5G TCIAM2672-25G
N-Methoxy-N-methylbenzamide ≥98.0% (by GC)
N-Methoxy-N-methylbenzamide
Formule: C₉H₁₁NO₂
Kookpunt: 112°C
MDL Nummer: MFCD00075320
CAS nummer: 6919-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34d001a229e19a69aa44e952004f68161fbd81e1&vid=34d001a229e19a69aa44e952004f68161fbd81e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.1050 to 1.1090
Refractive index n20/D 1.5320 to 1.5360


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR