Malotilate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (1,3-Dithiol-2-ylidene)malonic acid Diisopropyl ester, Diisopropyl (1,3-Dithiol-2-ylidene)malonate, Diisopropyl 2-(1,3-dithiol-2-ylidene)malonate

M2674-1G M2674-5G
TCIAM2674-1GEA 63.8 EUR
TCIAM2674-1G TCIAM2674-5G
Malotilate ≥98.0% (by GC)
Malotilate
Formule: C₁₂H₁₆O₄S₂
MW: 288.38 g/mol
MDL Nummer: MFCD00867646
CAS nummer: 59937-28-9
EINECS: 261-987-3
Merck Index: 14,05712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbf1d02d99939e4281bc80a56d3ee197c1a82506&vid=cbf1d02d99939e4281bc80a56d3ee197c1a82506&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR