Methyl-4-amino-3-chlorobenzoate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2698-1G M2698-5G
TCIAM2698-1GEA 33 EUR
TCIAM2698-1G TCIAM2698-5G
Methyl-4-amino-3-chlorobenzoate ≥97.0% (by GC)
Methyl-4-amino-3-chlorobenzoate
Formule: C₈H₈ClNO₂
MDL Nummer: MFCD01320686
CAS nummer: 84228-44-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff911fdaa0014dc990741424b03cb80e393449&vid=5fff911fdaa0014dc990741424b03cb80e393449&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR