Methyl-2,5-dichloronicotinate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M2752-200MG M2752-1G
TCIAM2752-200MGEA 23.1 EUR
TCIAM2752-200MG TCIAM2752-1G
Methyl-2,5-dichloronicotinate ≥98.0% (by GC)
Methyl-2,5-dichloronicotinate
Formule: C₇H₅Cl₂NO₂
MDL Nummer: MFCD03844857
CAS nummer: 67754-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc27d9bd232b8e24be9f7867aaae33964118c73b&vid=fc27d9bd232b8e24be9f7867aaae33964118c73b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C