4-(alpha-Methylbenzyl)resorcinol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-(α-Methylbenzyl)resorcinol, 4-(1-Phenylethyl)resorcinol, 4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcino
M2754-1G M2754-5G
TCIAM2754-1GEA 37.4 EUR
TCIAM2754-1G TCIAM2754-5G
4-(alpha-Methylbenzyl)resorcinol ≥98.0% (by GC)
4-(alpha-Methylbenzyl)resorcinol
Formule: C₁₄H₁₄O₂
MW: 214.26 g/mol
Kookpunt: 244°C
MDL Nummer: MFCD21648466
CAS nummer: 85-27-8
EINECS: 480-070-0
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C