4-Methyl-2,5-diphenylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2826-1G M2826-5G
TCIAM2826-1GEA 104 EUR
TCIAM2826-1G TCIAM2826-5G
4-Methyl-2,5-diphenylpyridine ≥98.0% (by GC)
4-Methyl-2,5-diphenylpyridine
Formule: C₁₈H₁₅N
MDL Nummer: MFCD13152024
CAS nummer: 156021-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b8192d74b417e9e55b239e8f02fd6a3d902b6f&vid=d8b8192d74b417e9e55b239e8f02fd6a3d902b6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR