5-Methyl-3,4-diphenylisoxazole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2830-1G
M2830-5G
TCIAM2830-1GEA
22
EUR
TCIAM2830-1G
TCIAM2830-5G
5-Methyl-3,4-diphenylisoxazole ≥98.0% (by GC)
5-Methyl-3,4-diphenylisoxazole
Formule:
C₁₆H₁₃NO |
MDL Nummer:
MFCD18449672 CAS nummer: 37928-17-9 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...