2-Methyl-4-phenylthiazol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2892-1G M2892-5G
TCIAM2892-1GEA 64.9 EUR
TCIAM2892-1G TCIAM2892-5G
2-Methyl-4-phenylthiazol ≥98.0% (by GC)
2-Methyl-4-phenylthiazol
Formule: C₁₀H₉NS
Kookpunt: 284°C
MDL Nummer: MFCD00489998
CAS nummer: 1826-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=706d2bdd1b83ec60ff2cd62cff9c57b36c920a51&vid=706d2bdd1b83ec60ff2cd62cff9c57b36c920a51&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C