Methyl 2-(2,6-Dichloroanilino)phenylacetate ≥97.0% (by GC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-(2,6-Dichloroanilino)phenylacetic acid methyl ester, Diclofenac methyl ester
M2903-200MG
M2903-1G
TCIAM2903-200MGEA
68.2
EUR
TCIAM2903-200MG
TCIAM2903-1G
Methyl 2-(2,6-Dichloroanilino)phenylacetate ≥97.0% (by GC, total nitrogen)
Methyl 2-(2,6-Dichloroanilino)phenylacetate
Formule:
C₁₅H₁₃Cl₂NO₂ MW: 310.17 g/mol Smeltpunt: 102°C |
MDL Nummer:
MFCD08273811 CAS nummer: 15307-78-5 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...