3-Methyl-4-nitrobenzyl alcohol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(3-Methyl-4-nitrophenyl)methanol, 5-(Hydroxymethyl)-2-nitrotoluene
M2915-1G
M2915-5G
TCIAM2915-1GEA
27.5
EUR
TCIAM2915-1G
TCIAM2915-5G
3-Methyl-4-nitrobenzyl alcohol ≥98.0% (by GC)
3-Methyl-4-nitrobenzyl alcohol
Formule:
C₈H₉NO₃ |
MDL Nummer:
MFCD00007171 CAS nummer: 80866-75-7 EINECS: 279-578-3 |
|
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...