Dimethyl-3-thiaadipate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2922-1G
M2922-5G
TCIAM2922-1GEA
26.4
EUR
TCIAM2922-1G
TCIAM2922-5G
Dimethyl-3-thiaadipate ≥98.0% (by GC)
Dimethyl-3-thiaadipate
Formule:
C₇H₁₂O₄S Kookpunt: 117°C Vlampunt: 54°C |
MDL Nummer:
MFCD00043633 CAS nummer: 7400-45-5 EINECS: 231-002-1 UN: 3272 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...