4-(2-Methoxyphenyl)butyric acid ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2926-1G
M2926-5G
TCIAM2926-1GEA
92.4
EUR
TCIAM2926-1G
TCIAM2926-5G
4-(2-Methoxyphenyl)butyric acid ≥97.0% (by GC, titration analysis)
4-(2-Methoxyphenyl)butyric acid
Formule:
C₁₁H₁₄O₃ Kookpunt: 160°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00796491 CAS nummer: 33209-75-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...