5-Methyl-1,3-benzenediacetonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
3,5-Bis(cyanomethyl)toluene, 2,2'-(5-Methyl-1,3-phenylene)diacetonitrile
M2934-5G
M2934-25G
TCIAM2934-5GEA
181
EUR
TCIAM2934-5G
TCIAM2934-25G
5-Methyl-1,3-benzenediacetonitrile ≥98.0% (by GC)
5-Methyl-1,3-benzenediacetonitrile
Formule:
C₁₁H₁₀N₂ Kookpunt: 225°C |
MDL Nummer:
MFCD07782113 CAS nummer: 120511-74-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...