Methyl-4-bromo-2-(bromomethyl)benzoate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Methyl 4-bromo-2-bromomethylbenzoate
M2941-200MG
M2941-1G
TCIAM2941-200MGEA
36.3
EUR
TCIAM2941-200MG
TCIAM2941-1G
Methyl-4-bromo-2-(bromomethyl)benzoate ≥97.0% (by GC)
Methyl-4-bromo-2-(bromomethyl)benzoate
Formule:
C₉H₈Br₂O₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD04114319 CAS nummer: 78471-43-9 UN: 3261 ADR: 8,III |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...