Methyl 5-Chlorothiophene-2-carboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2942-1G M2942-5G
TCIAM2942-1GEA 48.4 EUR
TCIAM2942-1G TCIAM2942-5G
Methyl 5-Chlorothiophene-2-carboxylate ≥98.0% (by GC)
Methyl 5-Chlorothiophene-2-carboxylate
Formule: C₆H₅ClO₂S
Kookpunt: 226°C
Smeltpunt: 18°C
MDL Nummer: MFCD06203698
CAS nummer: 35475-03-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR