5-Methylbenzo[b]thiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2974-1G M2974-5G
TCIAM2974-1GEA 88 EUR
TCIAM2974-1G TCIAM2974-5G
5-Methylbenzo[b]thiophene ≥98.0% (by GC)
5-Methylbenzo[b]thiophene
Formule: C₉H₈S
Kookpunt: 67°C
MDL Nummer: MFCD00052509
CAS nummer: 14315-14-1
EINECS: 238-256-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=020b1cd4c9dca847e9f037cf8277754fbe3abcc4&vid=020b1cd4c9dca847e9f037cf8277754fbe3abcc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR