2-Methyldibenzo[f,h]quinoxaline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M3005-200MG M3005-1G
TCIAM3005-200MGEA 48.4 EUR
TCIAM3005-200MG TCIAM3005-1G
2-Methyldibenzo[f,h]quinoxaline ≥98.0% (by GC)
2-Methyldibenzo[f,h]quinoxaline
Formule: C₁₇H₁₂N₂
MDL Nummer: MFCD16038624
CAS nummer: 536753-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fc66802453cb1c163a98b7cf0a2c31a68acb72e&vid=8fc66802453cb1c163a98b7cf0a2c31a68acb72e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR