Methyl-2-iodo-5-methylbenzoate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M3019-1G M3019-5G
TCIAM3019-1GEA 27.5 EUR
TCIAM3019-1G TCIAM3019-5G
Methyl-2-iodo-5-methylbenzoate ≥98.0% (by GC)
Methyl-2-iodo-5-methylbenzoate
Formule: C₉H₉IO₂
Kookpunt: 105°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12198129
CAS nummer: 103440-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16deb4fed0aa458951a4bc88595a66b605a4c6be&vid=16deb4fed0aa458951a4bc88595a66b605a4c6be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %