1-Methyl-3-propylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Methyl-3-propylimidazolium Tetrafluoroborate
M3036-5G
M3036-25G
TCIAM3036-5GEA
24.2
EUR
TCIAM3036-5G
TCIAM3036-25G
1-Methyl-3-propylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Methyl-3-propylimidazolium tetrafluoroborate
Formule:
C₇H₁₃BF₄N₂ Smeltpunt: 17°C |
MDL Nummer:
MFCD14156016 CAS nummer: 244193-48-4 UN: 1760 ADR: 8,II |
|
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...