1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide ≥97.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulphonyl)imide
M3039-5G
M3039-25G
TCIAM3039-5GEA
81.4
EUR
TCIAM3039-5G
TCIAM3039-25G
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide ≥97.0% (by HPLC)
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide
Formule:
C₁₄H₂₃F₆N₃O₄S₂ MW: 475.48 g/mol |
MDL Nummer:
MFCD08458946 CAS nummer: 178631-04-4 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...