Methyl 1-Formylcyclopropane-1-carboxylate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M3045-1G M3045-5G
TCIAM3045-1GEA 126 EUR
TCIAM3045-1G TCIAM3045-5G
Methyl 1-Formylcyclopropane-1-carboxylate ≥95.0% (by GC)
Methyl 1-Formylcyclopropane-1-carboxylate
Formule: C₆H₈O₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD15526934
CAS nummer: 88157-41-9
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR