3-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M3065-1G M3065-5G
TCIAM3065-1GEA 114 EUR
TCIAM3065-1G TCIAM3065-5G
3-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)
3-Methoxy-N-(4-methoxyphenyl)aniline
Formule: C₁₄H₁₅NO₂
Kookpunt: 195°C
CAS nummer: 3661-49-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR