N-Methoxy-N-methylpropionamide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M3096-1G M3096-5G
TCIAM3096-1GEA 41.8 EUR
TCIAM3096-1G TCIAM3096-5G
N-Methoxy-N-methylpropionamide ≥97.0% (by GC)
N-Methoxy-N-methylpropionamide
Formule: C₅H₁₁NO₂
MDL Nummer: MFCD09833541
CAS nummer: 104863-65-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR