Methyl-3-bromoisonicotinate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M3103-1G M3103-5G
TCIAM3103-1GEA 19.8 EUR
TCIAM3103-1G TCIAM3103-5G
Methyl-3-bromoisonicotinate ≥98.0% (by GC)
Methyl-3-bromoisonicotinate
Formule: C₇H₆BrNO₂
Kookpunt: 134°C
MDL Nummer: MFCD03788226
CAS nummer: 59786-31-1
Order Now
Specification Test Results
Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %