SMER28 ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-Allyl-6-bromoquinazolin-4-amine
M3106-25MG
M3106-100MG
TCIAM3106-25MGEA
165
EUR
TCIAM3106-25MG
TCIAM3106-100MG
SMER28 ≥98.0% (by HPLC)
SMER28
Formule:
C₁₁H₁₀BrN₃ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD02166825 CAS nummer: 307538-42-7 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 15.00 to 16.70 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...