cvxIAA ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-(3-Methoxyphenyl)indole-3-acetic acid, 5-(3-MeOPh)-IAA, convex IAA
M3141-10MG
M3141-50MG
TCIAM3141-10MGEA
150
EUR
TCIAM3141-10MG
TCIAM3141-50MG
cvxIAA ≥98.0% (by HPLC)
cvxIAA
Formule:
C₁₇H₁₅NO₃ MW: 281.31 g/mol |
CAS nummer:
2127037-35-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...