4-(Methylsulphonyl)-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M3146-25G M3146-5G
TCIAM3146-25GEA 36.3 EUR
TCIAM3146-25G TCIAM3146-5G
4-(Methylsulphonyl)-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-(Methylsulphonyl)-2-nitrobenzoic acid
Formule: C₈H₇NO₆S
MDL Nummer: MFCD01941299
CAS nummer: 110964-79-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR