2-Methyl-1-tetralone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M3159-1G M3159-5G
TCIAM3159-1GEA 41.8 EUR
TCIAM3159-1G TCIAM3159-5G
2-Methyl-1-tetralone ≥98.0% (by GC)
2-Methyl-1-tetralone
Formule: C₁₁H₁₂O
Kookpunt: 131°C
Smeltpunt: 15°C
MDL Nummer: MFCD00001690
CAS nummer: 1590-08-5
EINECS: 216-461-8
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %