Mal-PEG2-acid ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M3203-1G
M3203-250MG
TCIAM3203-1GEA
281
EUR
TCIAM3203-1G
TCIAM3203-250MG
Mal-PEG2-acid ≥98.0% (by HPLC)
Mal-PEG2-acid
Formule:
C₁₁H₁₅NO₆ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD23726620 CAS nummer: 1374666-32-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 95.0 to 105.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...