N²-Methyl-5-(trifluoromethyl)pyridine-2,3-diamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Amino-2-(methylamino)-5-(trifluoromethyl)pyridine, N2-Methyl-5-(trifluoromethyl)pyridine-2,3-diamine

M3215-1G M3215-5G
TCIAM3215-1GEA 95.7 EUR
TCIAM3215-1G TCIAM3215-5G
N²-Methyl-5-(trifluoromethyl)pyridine-2,3-diamine ≥98.0% (by GC, titration analysis)
N²-Methyl-5-(trifluoromethyl)pyridine-2,3-diamine
Formule: C₇H₈F₃N₃
MDL Nummer: MFCD00068119
CAS nummer: 172648-55-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR