1-Methylcyclopentyl acrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

M3261-1G M3261-5G
TCIAM3261-1GEA 61.6 EUR
TCIAM3261-1G TCIAM3261-5G
1-Methylcyclopentyl acrylate ≥98.0% (by GC) stabilised
1-Methylcyclopentyl acrylate
Formule: C₉H₁₄O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD08704215
CAS nummer: 178889-49-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR