3-Methylnonane-2,4-dione ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M3277-1G
M3277-5G
TCIAM3277-1GEA
140
EUR
TCIAM3277-1G
TCIAM3277-5G
3-Methylnonane-2,4-dione ≥97.0% (by GC)
3-Methylnonane-2,4-dione
Formule:
C₁₀H₁₈O₂ Kookpunt: 59°C |
MDL Nummer:
MFCD08741451 CAS nummer: 113486-29-6 |
Specification Test Results
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...