4'-Nitro-4-dimethylaminoazobenzene ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0192-25G
N0192-5G
TCIAN0192-25GEA
271
EUR
TCIAN0192-25G
TCIAN0192-5G
4'-Nitro-4-dimethylaminoazobenzene ≥98.0% (by titrimetric analysis)
4'-Nitro-4-dimethylaminoazobenzene
Formule:
C₁₄H₁₄N₄O₂ |
MDL Nummer:
MFCD00059834 CAS nummer: 2491-74-9 |
|
Specification Test Results
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 230.0 to 234.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...