Nitromethaan 55% ∼8.7 mol/L in methanol

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

N0239-25ML N0239-500ML
TCIAN0239-25MLEA 17.6 EUR
TCIAN0239-25ML TCIAN0239-500ML
Nitromethaan 55% ∼8.7 mol/L in methanol
Nitromethaan
Formule: CH₃NO₂
MDL Nummer: MFCD00007400
CAS nummer: 75-52-5
EINECS: 200-876-6
UN: 1993
ADR: 3,II
Merck Index: 14,06611
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f828d6be0e34b3a0b357a9a031db4729dbe1fbb&vid=d30720f3f12ece19a1d9f6e3c9244ee49a815c97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Concentration (GC) 55.0 to 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR