N-Nitrosodibutylamine ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-Dibutylnitrosamine

N0375-25ML N0375-5ML
TCIAN0375-25MLEA 178 EUR
TCIAN0375-25ML TCIAN0375-5ML
N-Nitrosodibutylamine ≥97.0% (by GC)
N-Nitrosodibutylamine
Formule: C₈H₁₈N₂O
Kookpunt: 237°C
MDL Nummer: MFCD00013892
CAS nummer: 924-16-3
EINECS: 213-101-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR