Norethindrone ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Norethisterone

N0449-1G N0449-100MG
TCIAN0449-1GEA 110 EUR
TCIAN0449-1G TCIAN0449-100MG
Norethindrone ≥98.0% (by titrimetric analysis)
Norethindrone
Formule: C₂₀H₂₆O₂
MDL Nummer: MFCD00067596
CAS nummer: 68-22-4
EINECS: 200-681-6
Merck Index: 14,06697

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 205.0 to 208.0 deg-C
Specific rotation [a]20/D -32.0 to -37.0 deg (C=1, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR