N-Nitrosomorpholine ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
N0466-25G
N0466-5G
TCIAN0466-25GEA
476
EUR
TCIAN0466-25G
TCIAN0466-5G
N-Nitrosomorpholine ≥99.0% (by GC)
N-Nitrosomorpholine
Formule:
C₄H₈N₂O₂ Kookpunt: 225°C |
MDL Nummer:
MFCD00039710 CAS nummer: 59-89-2 UN: 2811 ADR: 6.1,III Merck Index: 14,06640 |
Specification Test Results
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...