4-Nitrocinnamyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

N0690-1G N0690-5G
TCIAN0690-1GEA 47.3 EUR
TCIAN0690-1G TCIAN0690-5G
4-Nitrocinnamyl alcohol ≥98.0% (by GC)
4-Nitrocinnamyl alcohol
Formule: C₉H₉NO₃
MW: 179.18 g/mol
MDL Nummer: MFCD00017045
CAS nummer: 1504-63-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR