Nicergoline ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0904-1G
N0904-100MG
TCIAN0904-1GEA
196
EUR
TCIAN0904-1G
TCIAN0904-100MG
Nicergoline ≥98.0% (by HPLC, titration analysis)
Nicergoline
Formule:
C₂₄H₂₆BrN₃O₃ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00869626 CAS nummer: 27848-84-6 EINECS: 248-694-6 Merck Index: 14,09496 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Specific rotation [a]20/D | +5.2 to +6.2 deg (C=5, EtOH (95vol%)) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...