(4-Nitrophenyl)urea ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0980-1G
N0980-5G
TCIAN0980-1GEA
139
EUR
TCIAN0980-1G
TCIAN0980-5G
(4-Nitrophenyl)urea ≥98.0% (by HPLC, total nitrogen)
(4-Nitrophenyl)urea
Formule:
C₇H₇N₃O₃ Smeltpunt: 217°C |
MDL Nummer:
MFCD09040599 CAS nummer: 556-10-5 Merck Index: 14,06626 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...