Nimesulide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

N0984-1G N0984-5G
TCIAN0984-1GEA 17.6 EUR
TCIAN0984-1G TCIAN0984-5G
Nimesulide ≥98.0% (by HPLC, titration analysis)
Nimesulide
Formule: C₁₃H₁₂N₂O₅S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00079470
CAS nummer: 51803-78-2
EINECS: 257-431-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,06548
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe6a6f00031b2030aa01dd003965346a415f98ab&vid=fe6a6f00031b2030aa01dd003965346a415f98ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR