9-Nitroanthracene ≥90.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N0987-1G
N0987-5G
TCIAN0987-1GEA
46.2
EUR
TCIAN0987-1G
TCIAN0987-5G
9-Nitroanthracene ≥90.0% (by GC)
9-Nitroanthracene
Formule:
C₁₄H₉NO₂ Kookpunt: 275°C |
MDL Nummer:
MFCD00001248 CAS nummer: 602-60-8 EINECS: 210-021-9 |
|
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 90.0 % |
Melting point | 142.0 to 147.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...