Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
[NH2Me2][{RuCl((S)-dm-segphos®)}2(mu-Cl)3]
N1024-200MG
N1024-1G
TCIAN1024-200MGEA
39.6
EUR
TCIAN1024-200MG
TCIAN1024-1G
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Formule:
C₉₄H₉₆Cl₅NO₈P₄Ru₂ MW: 1871.09 g/mol |
MDL Nummer:
MFCD09753038 CAS nummer: 944451-14-3 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...