Nefazodone hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N1030-200MG
N1030-1G
TCIAN1030-200MGEA
28.6
EUR
TCIAN1030-200MG
TCIAN1030-1G
Nefazodone hydrochloride ≥98.0% (by HPLC, titration analysis)
Nefazodone hydrochloride
Formule:
C₂₅H₃₂ClN₅O₂·HCl MW: 506.47 g/mol |
MDL Nummer:
MFCD00935760 CAS nummer: 82752-99-6 Merck Index: 14,06438 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 186.0 to 190.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...